Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD166 Monoclonal Antibody (200622)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA523930

Catalog No. PIMA523930


Only null left
Add to Cart

Description

Description

In Western blots, shows 100% cross-reactivity with recombinant human (rh) ALCAM and no cross-reactivity with rhBCAM, rhEpCAM, recombinant mouse (rm) MAdCAM-1, rhMCAM, rmNCAM-L1, rmOCAM, or rmTROP-2. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD166
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q61490
ALCAM
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527.
100 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry, Western Blot
200622
Unconjugated
ALCAM
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product
Rat
Protein A/G
RUO
11658
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.