Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD166 Monoclonal Antibody (A6A8), Huabio

Mouse Monoclonal Antibody

Supplier:  HUABIO RESEARCH INC HA600083

Encompass
Encompass_Preferred

Catalog No. 50-254-4510


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

CD166 Monoclonal antibody specifically detects CD166 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

CD166
Monoclonal
2 mg/mL
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Q13740
Mouse
Protein G
RUO
214
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
A6A8
Unconjugated
ALCAM
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product
Recombinant protein within human CD166 aa 51-250.
100 μL
Primary
Human
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.