Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD169 Monoclonal Antibody (HSn 7D2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA116891
Description
CD169 Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), Flow, ELISA, RIA, IV
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
Specifications
CD169 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
A7LCJ3, Q62230, Q9BZZ2 | |
SIGLEC1 | |
Full length hSn(d1-4)Fc protein (purified) (Human). | |
100 μL | |
Primary | |
Human, Mouse, Pig, Virus | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry | |
HSn 7D2 | |
Unconjugated | |
SIGLEC1 | |
Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN | |
Mouse | |
Protein G | |
RUO | |
20612, 397623, 6614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction