Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD169 (Siglec-1) Monoclonal Antibody (MOMA-1), FITC
GREENER_CHOICE

Catalog No. PIMA528189
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528189 Supplier Invitrogen™ Supplier No. MA528189
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rat anti Mouse CD169, clone MOMA-1 recognizes murine CD169, also known as sialoadhesin or Siglec-1. CD169 is a lectin-like receptor expressed by certain populations of macrophages including marginal zone metallophils of the spleen, subcapsular macrophages of lymph nodes and stromal macrophages in bone marrow (Morris et al. 1991).

The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD169 (Siglec-1)
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone MOMA-1
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene SIGLEC1
Gene Accession No. Q62230
Gene Alias Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN
Gene Symbols SIGLEC1
Host Species Rat
Immunogen Stromal (reticular) elements from mouse lymph nodes.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20612
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.