Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD169 (Siglec-1) Monoclonal Antibody (SER-4), APC-eFluor™ 780, eBioscience™, Invitrogen™

Catalog No. 47575580
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-575-580 25 μg
47-575-582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-575-580 Supplier Invitrogen™ Supplier No. 47575580
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The SER-4 monoclonal antibody reacts with mouse CD169, which is also known as Sialoadhesin and Siglec-1. CD169 is a 185-kDa member of the Siglec family of adhesion and endocytic receptors that recognize sialylated glycans. This receptor is expressed on a subset of macrophages that are found with high frequency in the spleen, liver, lymph node, and bone marrow. CD169 can also be detected on monocytes in the bone marrow and periphery. This receptor plays a role in cell-cell adhesion, as well as internalization of sialylated virus and bacteria. This endocytic pathway allows CD169+ macrophages to present lipid antigens to iNKT cells, thereby leading to their activation. Studies have also demonstrated that direct interaction of CD169 with regulatory T cells inhibits their proliferation. The SER-4 antibody has been reported to have antagonist activity. Applications Reported: This SER-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SER-4 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD169 (Siglec-1)
Applications Flow Cytometry
Classification Monoclonal
Clone SER-4
Concentration 0.2 mg/mL
Conjugate APC-eFluor 780
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene SIGLEC1
Gene Accession No. Q62230
Gene Alias Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN
Gene Symbols SIGLEC1
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20612
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.