Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD16b (FCGR3B) Monoclonal Antibody (2D2G5B9)
GREENER_CHOICE

Catalog No. PIMA530591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530591 Supplier Invitrogen™ Supplier No. MA530591
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CD16b/FCGR3B.

CD16 (FCGR3A) is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by NK cells and some T cells, and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells. Human CD16 is expressed in two forms - FCGR3A and FCGR3B. FCGR3A is associated with the FcepsilonRI-gamma subunit and is responsible for antibody-dependent NK cell cytotoxicity. Mast cell FCGR3A is associated, with FcepsilonRI-beta subunit. Besides IgG, FCGR3A can be triggered also by oligomeric IgE. FCGR3B is a GPI-linked monomeric receptor expressed on neutrophils and is involved in their activation and induction of a pro-adhesive phenotype. Diseases associated with CD16 dysfunction include immunodeficiency 20 and systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16b (FCGR3B)
Applications ELISA
Classification Monoclonal
Clone 2D2G5B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FCGR3B
Gene Accession No. O75015
Gene Alias CD16; CD16B; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor IIIb; FCG3; Fc-gamma receptor IIIb (CD 16); Fc-gamma RIII; fc-gamma RIIIb; Fc-gamma RIII-beta; FCGR3; FCGR3B; FcR-10; FCRIII; FCRIIIb; IGFR3; IgG Fc receptor III-1; low affinity immunoglobulin gamma Fc region receptor III-B
Gene Symbols FCGR3B
Host Species Mouse
Immunogen Recombinant Human CD16b protein (Met1-Ser200).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.