Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16b (FCGR3B) Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | CD16b (FCGR3B) |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543908
|
Invitrogen™
MA543908 |
20 μL |
Each for $213.50
|
|
|||||
PIMA543907
|
Invitrogen™
MA543907 |
100 μL |
Each for $522.50
|
|
|||||
Description
CD16 (FCGR3A) is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by NK cells and some T cells, and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells. Human CD16 is expressed in two forms – FCGR3A and FCGR3B. FCGR3A is associated with the FcepsilonRI-gamma subunit and is responsible for antibody-dependent NK cell cytotoxicity. Mast cell FCGR3A is associated, with FcepsilonRI-beta subunit. Besides IgG, FCGR3A can be triggered also by oligomeric IgE. FCGR3B is a GPI-linked monomeric receptor expressed on neutrophils and is involved in their activation and induction of a pro-adhesive phenotype. Diseases associated with CD16 dysfunction include immunodeficiency 20 and systemic lupus erythematosus.Specifications
CD16b (FCGR3B) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
CD16; CD16B; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor IIIb; FCG3; Fc-gamma receptor IIIb (CD 16); Fc-gamma RIII; fc-gamma RIIIb; Fc-gamma RIII-beta; FCGR3; FCGR3B; FcR-10; FCRIII; FCRIIIb; IGFR3; IgG Fc receptor III-1; low affinity immunoglobulin gamma Fc region receptor III-B | |
FCGR3B | |
IgG2a κ | |
Protein A/G | |
Antibody |
C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O75015 | |
2215 | |
Pro24-Ser203 (Accession O75015), with N-terminal His Tag. | |
Primary | |
-20°C | |
FCGR3B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title