Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD17 Antibody (MEM-68), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP150060
Description
CD17 Monoclonal specifically detects CD17 in Human samples. It is validated for Flow Cytometry.Specifications
CD17 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 2 ug/ml | |
Mouse | |
Ion exchange chromatography | |
RUO | |
The mouse monoclonal antibody MEM-68 recognizes CD17, a membrane lipid moiety lactosylceramide expressed on granulocytes, monocytes and platelets. | |
Store at 4C. Do not freeze. | |
IgM |
Flow Cytometry | |
MEM-68 | |
Unconjugated | |
and amp;#223;-D-galactosyl-(1-4)- and amp;#223;-D-glucosyl-(1-1 and #039;)-ceramide, Lactosylceramide | |
Pre-B cell line NALM-6 | |
0.1 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction