Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD17 Monoclonal Antibody (MEM-68)
GREENER_CHOICE

Catalog No. PIMA110118
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110118 Supplier Invitrogen™ Supplier No. MA110118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes CD17, an extracellular membrane lipid moiety lactosylceramide expressed on granulocytes, monocytes and platelets.

CD17, lactosylceramide, is an ubiquitous glycosphingolipid with uncharged disaccharide headgroup, highly enriched in lipid raft-derived structures. Besides playing a pivotal role in the biosynthesis of complex glycosphingilipids, lactosylceramide is involved in cell-cell and cell-matrix interactions and in signaling events linked to cell differentiation, development, apoptosis and oncogenesis. Lactosylceramide regulates integrin functions and production of nitric oxide. Its expression defines successive stages in the maturation of myeloid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD17
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-68
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 15mM sodium azide; pH 8
Gene Alias Lactosylceramide
Host Species Mouse
Immunogen Pre-B cell line NALM-6.
Purification Method Sequential chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.