Learn More
CD170 Mouse anti-Human , PE, Clone: 1A5/CD170 (also known as 1A5), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD170 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1A5/CD170 (also known as 1A5) |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | SIGLEC-5; SIGLEC5; CD33 antigen-like 2; CD33L2; obesity-binding protein 2; OBBP2 |
| Host Species | Mouse |
| Immunogen | Human Siglec-5 Recombinant Protein |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.