Learn More
CD170 (Siglec F) Monoclonal Antibody (1RNM44N), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$217.00 - $441.00
Specifications
Antigen | CD170 (Siglec F) |
---|---|
Clone | 1RNM44N |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
36-317-0280
![]() |
Invitrogen™
363170280 |
25 μg |
Each for $217.00
|
|
|||||
36-317-0282
![]() |
Invitrogen™
363170282 |
100 μg |
Each for $441.00
|
|
|||||
Description
This 1RNM44N mAb recognizes mouse CD170, also known as Siglec F. Siglec F is a cell surface lectin belonging to the Ig superfamily that binds glycoconjugates containing sialic acids that are commonly found on various cell types. The cytoplasmic domain of Siglec F contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that initiates intracellular signaling upon ligand binding. Siglec F is expressed mostly on eosinophils and alveolar macrophages and lower levels of this receptor have also been reported on immature myelomonocytic cells. Mouse Siglec F is a functional ortholog of human Siglec 8, however, unlike human Siglec 8, mouse Siglec F is not expressed on mast cells. This 1RNM44N antibody will recognize a formaldehyde-fixed epitope. Applications Reported: This 1RNM44N antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1RNM44N antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. BUV395 is a dye that emits at 395nm and is intended for use on cytometers equipped with an Ultraviolet (355nm) laser. When using two or more Super Bright, BV, BUV, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (SB-4401-42) or Brilliant Stain Buffer (00-4409-75) to minimize any non-specific polymer interactions. Excitation: 350nm; Emission: 395nm; Laser: Ultraviolet Laser.
SIGLEC5 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. This protein binds equally to alpha2,3-linked and alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. This molecule is expressed by monocytic/myeloid lineage cells. SIGLEC5 is found at high levels in peripheral blood leukocytes, spleen, bone marrow and at lower levels in lymph node, lung, appendix, placenta, pancreas and thymus. It is expressed by monocytes and neutrophils but absent from leukemic cell lines representing early stages of myelomonocytic differentiation.Specifications
CD170 (Siglec F) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD170; CD33 antigen-like 2; CD33L2; mSiglec-F; OB-binding protein 2; OBBP2; OB-BP2; Obesity-binding protein 2; sialic acid binding Ig like lectin 5; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin F; sialic acid-binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin F; sialic acid-binding immunoglobulin-like lectin 5; sialic acid-binding lectin Siglec-F; Siglec; SIGLEC5; siglec-5; Siglecf; Siglec-F | |
Siglecf | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Siglecf |
1RNM44N | |
Flow Cytometry | |
Brilliant Ultraviolet 395 | |
Rat | |
Mouse | |
Q920G3 | |
233186 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.