Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD170 (Siglec F) Monoclonal Antibody (1RNM44N), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Catalog No. 367170280
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-717-0280 25 μg
36-717-0282 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-717-0280 Supplier Invitrogen™ Supplier No. 367170280
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

This 1RNM44N monoclonal antibody recognizes mouse CD170, also known as Siglec F. Siglec F is a cell surface lectin belonging to the Ig superfamily that binds glycoconjugates containing sialic acids that are commonly found on various cell types. The cytoplasmic domain of Siglec F contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that initiates intracellular signaling upon ligand binding. Siglec F is expressed mostly on eosinophils and alveolar macrophages and lower levels of this receptor have also been reported on immature myelomonocytic cells. Mouse Siglec F is a functional ortholog of human Siglec 8, however, unlike human Siglec 8, mouse Siglec F is not expressed on mast cells. This 1RNM44N antibody will recognize a formaldehyde-fixed epitope. Applications Reported: This 1RNM44N antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1RNM44N antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal macrophages. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Brilliant Ultraviolet 737 is a tandem dye that emits at 732 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.

SIGLEC-F is a sialic acid-binding immunoglobulin-like lectin predominantly expressed on eosinophils in mice, where it serves as a functional paralog of human SIGLEC-8. It acts as an inhibitory receptor with ITIM motifs that recognize sialylated glycans and can induce eosinophil apoptosis, thereby helping regulate allergic inflammation and immune homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD170 (Siglec F)
Applications Flow Cytometry
Classification Monoclonal
Clone 1RNM44N
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 737
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Siglecf
Gene Accession No. Q920G3
Gene Alias SIGLEC-8
Gene Symbols Siglecf
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233186
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.