Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD170 (Siglec F) Monoclonal Antibody (1RNM44N), eBioscience™, Invitrogen™

Catalog No. 14170237
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-170-237 2 mg
14-170-295 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-170-237 Supplier Invitrogen™ Supplier No. 14170237
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This 1RNM44N MAb recognizes mouse CD170, also known as Siglec F. Siglec F is a cell surface lectin belonging to the Ig superfamily that binds glycoconjugates containing sialic acids that are commonly found on various cell types. The cytoplasmic domain of Siglec F contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that initiates intracellular signaling upon ligand binding. Siglec F is expressed mostly on eosinophils and alveolar macrophages and lower levels of this receptor have also been reported on immature myelomonocytic cells. Mouse Siglec F is a functional ortholog of human Siglec 8, however, unlike human Siglec 8, mouse Siglec F is not expressed on mast cells. This 1RNM44N antibody will recognize a formaldehyde-fixed epitope.

SIGLEC-F is a sialic acid-binding immunoglobulin-like lectin predominantly expressed on eosinophils in mice, where it serves as a functional paralog of human SIGLEC-8. It acts as an inhibitory receptor with ITIM motifs that recognize sialylated glycans and can induce eosinophil apoptosis, thereby helping regulate allergic inflammation and immune homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD170 (Siglec F)
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 1RNM44N
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Siglecf
Gene Accession No. Q920G3
Gene Alias SIGLEC-8
Gene Symbols Siglecf
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233186
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.