Learn More
Invitrogen™ CD171 Recombinant Rabbit Monoclonal Antibody (014), PE

Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD171 (L1CAM) (Met1-Glu1120) and conjugated with PE under optimum conditions, the unreacted PE was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
Specifications
Specifications
| Antigen | CD171 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 014 |
| Concentration | 100 μg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.5% BSA and 0.09% sodium azide |
| Gene | L1CAM |
| Gene Accession No. | P32004 |
| Gene Alias | antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.