Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD172a/b Mouse anti-Human, Brilliant Violet 650, Clone: SE5A5, BD Optibuild™

Catalog No. BDB743565 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743565 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743565 Supplier BD Biosciences Supplier No. 743565
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SE5A5 monoclonal antibody specifically binds to a common epitope on CD172a/ SIRPα (90 kDa) and CD172b/ SIRPβ 1 (50 kDa) . These transmembrane glycoproteins are members of the Signal Regulatory Protein (SIRP) family that, in turn, belongs to the Immunoglobulin superfamily. The SIRP family is comprised of two subgroups, SIRPα and SIRPβ that are distinguished by the presence (α) or absence (β) of a cytoplasmic domain containing two immunoreceptor tyrosine-based inhibition motifs (ITIM). CD172a/ SIRPα is expressed on CD34+ stem/progenitor cells, cardiomyocytes, monocytes, macrophages, granulocytes, dendritic cells, and in the central nervous system. It binds to CD47 and is implicated in mediating inhibitory signals via the ITIM/SHP-2 association. CD172b/ SIRPβ 1 does not possess a cytoplasmic domain but instead the transmembrane domain contains a positively-charged residue that can interact with another transmembrane protein, DAP-12/KARAP. DAP-12 has two immunoreceptor tyrosine-based activat ion motifs (ITAM) within its cytoplasmic domain that are thought to link CD172b to cellular activation signaling . CD172b is expressed on myeloid cells, including peripheral blood monocytes and granulocytes. It is not expressed on CD34+ cells. CD172a and CD172b have complementary role s in signal regulation and may work together in tuning certain cellular responses to stimuli.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD172a/b
Classification Monoclonal
Clone SE5A5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SIRP alpha/beta1, SIRPα/SIRPβ1, Signal Regulatory Protein α/β1
Host Species Mouse
Immunogen Human SIRP alpha extracellular domain Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.