Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD172a/b Mouse anti-Human, BUV395, Clone: SE5A5, BD Optibuild™

Catalog No. BDB743568 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743568 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743568 Supplier BD Biosciences Supplier No. 743568
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SE5A5 monoclonal antibody specifically binds to a common epitope on CD172a/ SIRPα (90 kDa) and CD172b/ SIRPβ 1 (50 kDa) . These transmembrane glycoproteins are members of the Signal Regulatory Protein (SIRP) family that, in turn, belongs to the Immunoglobulin superfamily. The SIRP family is comprised of two subgroups, SIRPα and SIRPβ that are distinguished by the presence (α) or absence (β) of a cytoplasmic domain containing two immunoreceptor tyrosine-based inhibition motifs (ITIM). CD172a/ SIRPα is expressed on CD34+ stem/progenitor cells, cardiomyocytes, monocytes, macrophages, granulocytes, dendritic cells, and in the central nervous system. It binds to CD47 and is implicated in mediating inhibitory signals via the ITIM/SHP-2 association. CD172b/ SIRPβ 1 does not possess a cytoplasmic domain but instead the transmembrane domain contains a positively-charged residue that can interact with another transmembrane protein, DAP-12/KARAP. DAP-12 has two immunoreceptor tyrosine-based activat ion motifs (ITAM) within its cytoplasmic domain that are thought to link CD172b to cellular activation signaling . CD172b is expressed on myeloid cells, including peripheral blood monocytes and granulocytes. It is not expressed on CD34+ cells. CD172a and CD172b have complementary role s in signal regulation and may work together in tuning certain cellular responses to stimuli.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD172a/b
Classification Monoclonal
Clone SE5A5
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SIRP alpha/beta1, SIRPα/SIRPβ1, Signal Regulatory Protein α/β1
Host Species Mouse
Immunogen Human SIRP alpha extracellular domain Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.