Learn More
Invitrogen™ CD172a (SIRP alpha) Monoclonal Antibody (OX41), APC, eBioscience™

Description
Description: The monoclonal antibody OX41 reacts with rat CD172a, which is also known as signal-regulatory protein alpha (SIRPa). SIRP family members are transmembrane glycoproteins with 3 Ig-like extracellular domains and a cytoplasmic tail containing 2 immunocreceptor tyrosine-based inhibition motifs. CD172a is expressed by monocytes, macrophages, granulocytes, dendritic cells, and neurons. SIRPa association with CD47 inhibits phagocytosis of CD47 presenting cells. Studies suggest that SIRPa ligation prevents phagocytosis by inactivating paxillin and cofilin, proteins involved in cytoskeleton remodeling. Applications Reported: This OX41 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX41 antibody has been tested by flow cytometric analysis of rat lysed whole blood. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD172a (SIRP alpha) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OX41 |
| Concentration | 0.2 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | SIRPA |
| Gene Accession No. | P97710 |
| Gene Alias | AI835480; Bit; brain Ig-like molecule with tyrosine-based activation motifs; Brain immunoglobulin like protein with tyrosine - based activation motifs; brain immunological-like with tyrosine-based motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172; CD172 alpha; CD172 antigen-like family member A; CD172A; Inhibitory receptor SHPS-1; macrophage fusion receptor; Macrophage membrane protein MFP150; MFR; mSIRP-alpha1; Myd1; MYD-1; myD-1 antigen; p84; Protein tyrosine phosphatase non-receptor type substrate 1 (SHP substrate 1); protein tyrosine phosphatase, non-receptor type substrate 1; Protein tyrosine phosphatase, non-receptor type substrate 1 (SHP substrate 1); Ptpns1; SHP substrate 1; SHP-1; SHPS1; SHPS-1; signal regulatory protein alpha; signal-regulatory protein alpha; signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; SIRP; Sirpa; SIRPalpha; Sirp-alpha-1; SIRPalpha2; Sirp-alpha-2; Sirp-alpha-3; swc3; swine workshop cluster 3 antigen; swine workshop cluster 3 antigen precursor; tyrosine phosphatase SHP substrate 1; tyrosine-protein phosphatase non-receptor type substrate 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.