Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD173 Antibody (A51-B/A6), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314046AF750
This item is not returnable.
View return policy
Description
CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CD173 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| H2, Type 2 H blood group antigen | |
| Human breast cancer MCF-7 cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgA |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| A51-B/A6 | |
| 50mM Sodium Borate | |
| Mouse | |
| PEG purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction