Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD173 Antibody (BRIC231), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062120AF750
Description
CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry.Specifications
CD173 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes | |
Store at 4°C in the dark. |
Flow Cytometry | |
BRIC231 | |
Flow Cytometry | |
H2, Type 2 H blood group antigen | |
HEL cell line | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction