Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD173 Antibody (BRIC231), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | CD173 |
---|---|
Clone | BRIC231 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000, Haemagglutination |
Applications | Hemagglutination Assay, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017432
![]() |
Novus Biologicals
NB100621200.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10062120
![]() |
Novus Biologicals
NB10062120 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry, Haemagglutination, Immunofluorescence.Specifications
CD173 | |
1 mg/mL | |
Hemagglutination Assay, Flow Cytometry | |
Unconjugated | |
RUO | |
HEL cell line | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
BRIC231 | |
Flow Cytometry 1:10-1:1000, Haemagglutination | |
Monoclonal | |
Mouse | |
Human | |
IgG1 | |
Affinity Purified | |
NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title