Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD173 and CD174 Mouse anti-Human, Clone: BRIC 231, MilliporeSigma™

Catalog No. MABS1927MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1927MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1927MI Supplier MilliporeSigma Supplier No. MABS1927
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD173 CD174, clone BRIC 231 is a mouse monoclonal antibody that detects CD173/CD174 in human and is tested for use in Flow cytometry and Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD173 and CD174
Applications Immunocytochemistry, Flow Cytometry
Classification Monoclonal
Clone BRIC 231
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Whole HEL cells
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002024
Test Specificity Clone BRIC231 detects CD173/CD174 in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.