Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD173 Monoclonal Antibody (BRIC231)
GREENER_CHOICE

Catalog No. PIMA182022
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA182022 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA182022 Supplier Invitrogen™ Supplier No. MA182022
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD173 antibody, clone BRIC231 recognizes human type 2 H blood group antigen, also known as CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes.

CD173 is also known as human Type 2 H blood group antigen. Active H substances are expressed by many cells and tissues, including erythrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD173
Applications Flow Cytometry, Hemagglutination Assay, Immunocytochemistry
Classification Monoclonal
Clone BRIC231
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation tris with glycine and <0.1% sodium azide; pH 7.4
Gene Alias H2; Type 2 H blood group antigen
Host Species Mouse
Immunogen HEL cell line.
Purification Method purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.