Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD173 Mouse anti-Human, Clone: A51-B/A6, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | CD173 |
---|---|
Clone | A51-B/A6 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry 1-2 ug/ml, Immunofluorescence 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CD173 | |
0.2 mg/ml | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P16442 | |
Human breast cancer MCF-7 cells | |
Primary | |
Store at 4°C. |
A51-B/A6 | |
Immunohistochemistry 1-2 ug/ml, Immunofluorescence 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
H2, Type 2 H blood group antigen | |
IgA | |
PEG purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title