Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD175 Monoclonal Antibody (BRIC111)
GREENER_CHOICE

Catalog No. MA180055
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA180055 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA180055 Supplier Invitrogen™ Supplier No. MA180055
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-80055 recognizes Tn antigen on glycophorin A and glycophorin B in human erythrocytes. A suggested positive control for immunohistochemical applications is breast carcinoma.

Tn, recently designated CD175, is a cryptantigen associated with polyagglutination. It is not expressed on normal haemopoietic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD175
Applications ELISA, Hemagglutination Assay, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BRIC111
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Tris with glycine and <0.1% sodium azide; pH 7.4
Gene Alias CD175s
Host Species Mouse
Immunogen Human Tn red blood cells.
Purification Method Purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.