Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD176 Monoclonal Antibody (A78-G/A7)
GREENER_CHOICE

Catalog No. MA134862
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA134862 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA134862 Supplier Invitrogen™ Supplier No. MA134862
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive controls for this product are KG1 cells and human colorectal carcinoma tissues.

The Thomsen-Fridenreich antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. It is considered as a pan-carcinoma marker. This gene is a member of the mucin family and encodes a membrane bound, glycosylated phosphoprotein. The protein is anchored to the apical surface of many epithelia by a transmembrane domain, with the degree of glycosylation varying with cell type. It also includes a 20 aa variable number tandem repeat (VNTR) domain, with the number of repeats varying from 20 to 120 in different individuals. The protein serves a protective function by binding to pathogens and also functions in a cell signaling capacity. Overexpression, aberrant intracellular localization, and changes in glycosylation of this protein have been associated with carcinomas. Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length nature of only some has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD176
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone A78-G/A7
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
Gene Tf
Gene Accession No. P02787, P12346
Gene Alias TFA
Gene Symbols Tf
Host Species Mouse
Immunogen Neuraminidase-treated human red blood cells.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24825, 7018
Target Species Human, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.