Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD177 Antibody (021), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290285AF350
Description
CD177 Monoclonal specifically detects CD177 in Human samples. It is validated for ELISA.Specifications
CD177 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
57126 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
21 | |
ELISA | |
CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1NB1 glycoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD177 (AAH29167.1; Met1-Gly407). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction