Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD177 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CD177 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CD177 Polyclonal specifically detects CD177 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD177 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1NB1 glycoprotein | |
CD177 | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
57126 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CQFGTVQHVWKVSDLPRQWTPKNTSCDSGLGCQDTLMLIESGPQVSLVLSKGCTEAKDQEPRVTEHRMGPGLSLISYTFVCRQEDFCNNLVNSLP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title