Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD177 Monoclonal Antibody (MEM-166 (MEM166)), eBioscience™
GREENER_CHOICE

Catalog No. 5012942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-129-42 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-129-42 Supplier Invitrogen™ Supplier No. 14177982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MEM-166 monoclonal antibody reacts with human CD177, a 58-64 kDa GPI-anchored glycoprotein. The human CD177 locus codes for at least two alleles, NB1 and PRV-1, which differ by only 4 amino acids. CD177 is expressed on granulocytes, early erythroblasts, megakaryocytes, promyelocytes and myelocytes. CD177 mRNA levels are elevated in several different myeloproliferative disorders including polycythemia vera, and antibodies to CD177 are involved in neonatal alloimmune neutropenia, autoimmune drug inducedneutropenia, and graft failure following marrow transplantation. Applications Reported: This MEM-166 (MEM166) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB) (non-reducing conditions). Applications Tested: This MEM-166 (MEM166) antibody has been tested by flow cytometric analysis of normal human peripheral blood. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD177
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM-166 (MEM166)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD177
Gene Accession No. Q8N6Q3
Gene Alias 1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181
Gene Symbols CD177
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57126
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.