Learn More
Invitrogen™ CD177 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593050
Description
Immunogen sequence: VSDLPRQWTP KNTSCDSGLG CQDTLMLIES GPQVSLVLSK GCTEAKDQEP RVTEHRMGPG LSLISYTFVC RQEDFCNNLV NSLPLWAPQP PADPGSLRCP VCLSMEGCLE GTTEEICPKG TTHCYDGLLR LRGGGIFSNL RVQGCMPQPV CNLLNGTQEI GPVGMTENCD MKDFLTCHRG TTIMTHGNLA QEPTDWTTSN TEMCEVGQVC QETLLLLDVG LTSTLVGTKG CSTVGAQNSQ KTTIHSAPPG V; Positive Samples: BT-474; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).
Specifications
CD177 | |
Polyclonal | |
Unconjugated | |
CD177 | |
1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57126, 68891 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N6Q3, Q8R2S8 | |
CD177 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 35-285 of human CD177 (NP_0651392). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.