Learn More
CD178 (FasL) Monoclonal Antibody (MFL3), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Armenian Hamster Monoclonal Antibody
Supplier: Leinco Technologies Inc M351100MG
Description
Specificity: Clone MFL3 recognizes an epitope on mouse FasL. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
CD178 (FasL) Monoclonal antibody specifically detects CD178 (FasL) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Inhibition AssaysSpecifications
| CD178 (FasL) | |
| Monoclonal | |
| ≥5.0 mg/mL | |
| PBS with no preservative; pH 7.2-7.4 | |
| P41047 | |
| Armenian Hamster | |
| Protein A/G | |
| RUO | |
| 14103 | |
| 4°C short term, -80°C long term | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunocytochemistry, Inhibition Assays | |
| MFL3 | |
| Functional Grade | |
| Fasl | |
| ADAM10-processed FasL form; ALPS1B; APL; Apoptosis (APO 1) antigen ligand 1; apoptosis (APO-1) antigen ligand 1; Apoptosis (APO-1) antigen ligand 1 (Fas antigen ligand); apoptosis antigen ligand; APT1LG1; APTL; CD178; CD178 antigen; CD95 ligand; CD95 ligand protein;Generalized lymphoproliferative disease (Gld); Cd95l; CD95-L; CD95L protein; fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); Fasl; Fas-L; FasL ICD; FasL intracellular domain; Faslg; Fas-LG; generalized lymphoproliferative disease; gld; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; soluble form; SPA; SPPL2A-processed FasL form; TNFL; Tnfsf6; TNLG1A; Tumor necrosis factor (ligand) superfamily member 6; Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor (ligand) superfamily, member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor ligand; tumor necrosis factor ligand 1A; tumor necrosis factor ligand superfamily member 6; Tumor necrosis factor ligand superfamily member 6 (TNFL6 / TNFSF6); Tumor necrosis factor ligand superfamily member 6, membrane form; Tumor necrosis factor ligand superfamily member 6, soluble form | |
| B6 mouse FasL cDNA-transfected baby hamster kidney (B6 FasL/BHK) cells | |
| 100 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.