Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD178 Mouse anti-Human, BUV615, Clone: NOK-1, BD OptiBuild™

Catalog No. BDB752306 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752306 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752306 Supplier BD Biosciences Supplier No. 752306
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD178 Monoclonal antibody specifically detects CD178 in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD178
Applications Flow Cytometry
Classification Monoclonal
Clone NOK-1
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu CD178 BUV615 NOK-1 50 μg
Host Species Mouse
Immunogen Mouse T lymphoma cells (L5178Y) expressing human FasL
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD178 (Fas Ligand)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.