Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD179a Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563199
Description
Immunogen sequence: QPVLHQPPAM SSALGTTIRL TCTLRNDHDI GVYSVYWYQQ RPGHPPRFLL RYFSQSDKSQ GPQVPPRFSG SKDVARNRGY LSISELQPED EAMYYCAMGA RSSEKEERER EWEEEMEPT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.
The protein encoded by this gene belongs to the immunoglobulin superfamily and is expressed selectively at the early stages of B cell development, namely, in proB and early preB cells. This gene encodes the iota polypeptide chain that is associated with the Ig-mu chain to form a molecular complex which is expressed on the surface of pre-B cells. The complex is thought to regulate Ig gene rearrangements in the early steps of B-cell differentiation.
Specifications
CD179a | |
Polyclonal | |
Unconjugated | |
VPREB1 | |
CD179 antigen-like family member A; CD179a; IGI; IGLL1; IGVPB; Immunoglobulin iota chain; Immunoglobulin lambda Vpreb1 chain; immunoglobulin lambda-1 light chain; immunoglobulin lambda-like polypeptide 5; LOC100735142; pre-B lymphocyte 1; pre-B lymphocyte gene 1; Protein VPreB1; surrogate light chain; v(pre)B protein; VPREB; VpreB protein; Vpreb1; Vpreb-1; V-set pre-B cell surrogate light chain 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P12018 | |
VPREB1 | |
Recombinant protein corresponding to Human CD179a. Recombinant protein control fragment (Product #RP-91363). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction