Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD179a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103904 Supplier Invitrogen™ Supplier No. PA5103904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VPREB1.

The protein encoded by this gene belongs to the immunoglobulin superfamily and is expressed selectively at the early stages of B cell development, namely, in proB and early preB cells. This gene encodes the iota polypeptide chain that is associated with the Ig-mu chain to form a molecular complex which is expressed on the surface of pre-B cells. The complex is thought to regulate Ig gene rearrangements in the early steps of B-cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD179a
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VPREB1
Gene Accession No. P12018
Gene Alias CD179 antigen-like family member A; CD179a; IGI; IGLL1; IGVPB; Immunoglobulin iota chain; Immunoglobulin lambda Vpreb1 chain; immunoglobulin lambda-1 light chain; immunoglobulin lambda-like polypeptide 5; LOC100735142; pre-B lymphocyte 1; pre-B lymphocyte gene 1; Protein VPreB1; surrogate light chain; v(pre)B protein; VPREB; VpreB protein; Vpreb1; Vpreb-1; V-set pre-B cell surrogate light chain 1
Gene Symbols VPREB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human VPREB1(Accession P12018), corresponding to amino acid residues W56-Q106.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7441
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.