Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD179a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529714 Supplier Invitrogen™ Supplier No. PA529714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: K562, THP-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the immunoglobulin superfamily and is expressed selectively at the early stages of B cell development, namely, in proB and early preB cells. This gene encodes the iota polypeptide chain that is associated with the Ig-mu chain to form a molecular complex which is expressed on the surface of pre-B cells. The complex is thought to regulate Ig gene rearrangements in the early steps of B-cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD179a
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene VPREB1
Gene Accession No. P12018
Gene Alias CD179 antigen-like family member A; CD179a; IGI; IGLL1; IGVPB; Immunoglobulin iota chain; Immunoglobulin lambda Vpreb1 chain; immunoglobulin lambda-1 light chain; immunoglobulin lambda-like polypeptide 5; LOC100735142; pre-B lymphocyte 1; pre-B lymphocyte gene 1; Protein VPreB1; surrogate light chain; v(pre)B protein; VPREB; VpreB protein; Vpreb1; Vpreb-1; V-set pre-B cell surrogate light chain 1
Gene Symbols VPREB1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VpreB. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.