Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD179a (VpreB) Mouse anti-Human, BV421, Clone: HSL96, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566583
Description
CD179a (VpreB) Monoclonal antibody specifically detects CD179a (VpreB) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD179a (VpreB) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| HSL96 | |
| Brilliant Violet 421 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction