Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 (Integrin β2 chain) Rat anti-Mouse, BUV615, Clone: M18/2, BD OptiBuild™

Catalog No. BDB752359 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752359 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752359 Supplier BD Biosciences Supplier No. 752359
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD18 (Integrin β2 chain) Monoclonal antibody specifically detects CD18 (Integrin β2 chain) in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18 (Integrin β2 chain)
Applications Flow Cytometry
Classification Monoclonal
Clone M18/2
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms CD18 BUV615 M18/2 50 μg
Host Species Rat
Immunogen Mouse CTL glycoproteins
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD18 (Integrin β2 chain)
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.