Learn More
CD18 Monoclonal Antibody (3EB), Biotin, Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0183100TESTS


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD18 Monoclonal antibody specifically detects CD18 in Human samples. It is validated for Flow CytometrySpecifications
CD18 | |
Monoclonal | |
Biotin | |
ITGB2 | |
2E6; AI528527; antigen CD18; B-2 integrin; CD18; CD18 leukocyte adhesion molecule; CD18 subunit; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; complement receptor C3 subunit beta; integrin beta 2; integrin beta chain, beta 2; integrin beta-2; integrin subunit beta 2; integrin, beta 2; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; macrophage antigen 1 (mac-1) beta subunit); integrin, beta 2 (antigen CD18 subunit (p95), lymphocyte function-associated antigen 1, integrin B2); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGB2; LAD; LCAMB; Leu-CAM receptor; leukocyte cell adhesion molecule CD18; leukocyte surface protein; leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C; Lfa1; LFA-1; lymphocyte function associated antigen 1; MAC-1; Mac-1 beta; macrophage antigen 1 (mac-1) beta subunit); macrophage antigen-1 beta; MF17; MFI7 | |
Protein A/G | |
RUO | |
3689 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
3EB | |
PBS with 50% glycerol and 0.1% chloroacetamide | |
P05107 | |
Mouse | |
100Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.