Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Mouse, anti-Human, R718, Clone: L130, BD OptiBuild™

Catalog No. BDB752264 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752264 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752264 Supplier BD Biosciences Supplier No. 752264
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L130 monoclonal antibody specifically binds to CD18 which is also known as β2 integrin. CD18 is an ∼90-95 kDa type I transmembrane glycoprotein that is encoded by ITGB2. CD18 associates with the integrin α chains CD11a, CD11b, CD11c, or CD11d to form the LFA-1, Mac-1, p150,95, or αDβ2 heterodimers, respectively. CD18 is expressed on virtually all normal peripheral blood leucocytes. CD18 serves as a leucocyte adhesion receptor and signaling receptor. It is essential for many immune responses that require cell-to-cell contact, such as lymphocyte adhesion, natural killer (NK) cell and T-lymphocyte cytolysis, and T lymphocyte proliferation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Flow Cytometry
Classification Monoclonal
Clone L130
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.