Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Mouse, anti-Mouse, R718, Clone: C71/16, BD OptiBuild™

Catalog No. BDB752046 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752046 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752046 Supplier BD Biosciences Supplier No. 752046
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C71/16 monoclonal antibody specifically binds to mouse CD18 which is also known as the common β2 chain of LFA-1 (CD11a/CD18, αLβ2 integrin), Mac-1 (CD11b/CD18, αMβ2 integrin), and gp150, 95 (CD11c/CD18, αxβ2 integrin). CD18 is a type I transmembrane glycoprotein. Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with the three integrin α chains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. The β2 integrins are important mediators of leukocyte-endothelium interactions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Flow Cytometry
Classification Monoclonal
Clone C71/16
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.