Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Rat anti-Mouse, Brilliant Violet 421, Clone: M18/2, BD Optibuild™

Catalog No. BDB744597 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744597 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744597 Supplier BD Biosciences Supplier No. 744597
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M18/2 antibody specifically recognizes the commonβ2 chain of LFA-1 (CD11a/CD18,αLβ2 integrin), Mac-1 (CD11b/CD18,αMβ2 integrin), and gp150, 95 (CD11c/CD18,αXβ2 integrin). Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with the three integrinαchains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. Theβ2 integrins are important mediators of leukocyte-endothelium interactions. It has been reported that M18/2 antibody blocks in vivo /metastasis of the LB lymphoma to the spleen and that it blocks in vitro formation of aggregates of LB cells and splenocytes. However, other reports indicate that mAb M18/2 has no effect on CTLmediated killing, adherence of C3bi-sensitized erythrocytes to Mac-1, antigen-specific binding of T cells to antigen-producing cells, or rejection of cardiac allografts. Recent in vitro studies indicate that M18/2 antibody stimulates adhesion of Mac-1 to its ligands C3bi and ICAM-1, and it stimulates adhesion of LFA-1 to ICAM-1, but it has no effect upon the interactions of LFA-1 with ICAM-2 nor ICAM-3.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Classification Monoclonal
Clone M18/2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd18, ITGB2, Integrin β2 chain
Host Species Rat
Immunogen Mouse CTL glycoproteins
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.