Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Rat anti-Mouse, Brilliant Violet 650, Clone: M18/2, BD Optibuild™

Catalog No. BDB744600 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744600 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744600 Supplier BD Biosciences Supplier No. 744600
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M18/2 antibody specifically recognizes the common β 2 chain of LFA-1 (CD11a/CD18, α Lβ 2 integrin), Mac-1 (CD11b/CD18, α Mβ 2 integrin), and gp150, 95 (CD11c/CD18, α Xβ 2 integrin). Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with the three integrin α chains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. The β 2 integrins are important mediators of leukocyte-endothelium interactions. It has been reported that M18/2 antibody blocks in vivo metastasis of the LB lymphoma to the spleen and that it blocksi in vitro formation of aggregates of LB cells and splenocytes. However, other reports indicate that mAb M18/2 has no effect on CTLmediated killing, adherence of C3bi-sensitized erythrocytes to Mac-1, antigen-specific binding of T cells to antigen-producing cells, or rejection of cardiac allografts. Recent in vitro studies indicate that M18/2 antibody stimulates adhesion of Mac-1 to its ligands C3bi and ICAM-1, and it stimulates adhesion of LFA-1 to ICAM-1, but it has no effect upon the interactions of LFA-1 with ICAM-2 nor ICAM-3.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Classification Monoclonal
Clone M18/2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd18, ITGB2, Integrin β2 chain
Host Species Rat
Immunogen Mouse CTL glycoproteins
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.