Learn More
CD18 Rat anti-Mouse, BUV395, Clone: M18/2, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 744603
Description
The M18/2 antibody specifically recognizes the commonβ2 chain of LFA-1 (CD11a/CD18,αLβ2 integrin), Mac-1 (CD11b/CD18,αMβ2 integrin), and gp150, 95 (CD11c/CD18,αXβ2 integrin). Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with the three integrinαchains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. Theβ2 integrins are important mediators of leukocyte-endothelium interactions. It has been reported that M18/2 antibody blocks in vivo /metastasis of the LB lymphoma to the spleen and that it blocks in vitro formation of aggregates of LB cells and splenocytes. However, other reports indicate that mAb M18/2 has no effect on CTLmediated killing, adherence of C3bi-sensitized erythrocytes to Mac-1, antigen-specific binding of T cells to antigen-producing cells, or rejection of cardiac allografts. Recent in vitro studies indicate that M18/2 antibody stimulates adhesion of Mac-1 to its ligands C3bi and ICAM-1, and it stimulates adhesion of LFA-1 to ICAM-1, but it has no effect upon the interactions of LFA-1 with ICAM-2 nor ICAM-3.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
CD18 | |
M18/2 | |
Brilliant Ultraviolet 395 | |
Cd18, ITGB2, Integrin β2 chain | |
Mouse CTL glycoproteins | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.