Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Mouse anti-Human, Brilliant Violet 786, Clone: G28-8, BD Optibuild™

Catalog No. BDB743629 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743629 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743629 Supplier BD Biosciences Supplier No. 743629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G28-8 monoclonal antibody specifically recognizes RP105/Bgp95, a 95-105 kDa type I membrane protein consisting of extracellular leucine-rich repeats and a short cytoplasmic domain. It is expressed on mantle zone B cells, but weakly on germinal center B cells. RP105/Bgp95 is also expressed on peripheral blood monocytes, dendritic cells, and a subset of peripheral blood lymphocytes. The extracellular domain associates with a molecule called MD-1 to form a cell surface receptor complex RP105/Bgp95/MD- 1. This receptor belongs to the family of toll-like receptors (TLR). Studies show that RP105/Bgp95/MD-1, working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS). Reports on functional studies show that G28-8 monoclonal antibody can induce a G0 to G1 cell cycle transition and was synergistic with PMA, anti-μ, or anti-CD40 in inducing proliferation of resting B cells.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD180
Classification Monoclonal
Clone G28-8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias RP105, Lymphocyte antigen 64, LY64, Bgp95
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.