Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Mouse anti-Human, BUV395, Clone: G28-8, BD Optibuild™

Catalog No. BDB743630 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743630 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743630 Supplier BD Biosciences Supplier No. 743630
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G28-8 monoclonal antibody specifically recognizes RP105/Bgp95, a 95-105 kDa type I membrane protein consisting of extracellular leucine-rich repeats and a short cytoplasmic domain. It is expressed on mantle zone B cells, but weakly on germinal center B cells. RP105/Bgp95 is also expressed on peripheral blood monocytes, dendritic cells, and a subset of peripheral blood lymphocytes. The extracellular domain associates with a molecule called MD-1 to form a cell surface receptor complex RP105/Bgp95/MD- 1. This receptor belongs to the family of toll-like receptors (TLR). Studies show that RP105/Bgp95/MD-1, working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS). Reports on functional studies show that G28-8 monoclonal antibody can induce a G0 to G1 cell cycle transition and was synergistic with PMA, anti-μ, or anti-CD40 in inducing proliferation of resting B cells.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD180
Classification Monoclonal
Clone G28-8
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias RP105, Lymphocyte antigen 64, LY64, Bgp95
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.