Learn More
Invitrogen™ CD180 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110673
Description
Immunogen sequence: DFQNNAIHYI SREDMRSLEQ AINLSLNFNG NNVKGIELGA FDSTVFQSLN FGGTPNLSVI FNGLQNSTTQ SLWLGTFEDI DDEDISSAML KGLCEMSVES LNLQEHRFSD ISSTTFQCFT QLQELDLTAT HLKGLPSGMK GLNLLKKL Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 74%.
CD180 (RP105, Bgp95, LY64) is a type I membrane glycoprotein of Toll-like receptor (TLR) family. CD180 cytoplasmic tail is short and unlike the TLRs, it lacks the TIR domain. CD180 expression is dependent on the coexpression of its helper molecule, MD-1, and mirrors that of TLR4 on antigen-presenting cells. CD180 regulates recognition of LPS and signaling in B cells, via interacting directly with the TLR4 signaling complex, inhibiting its ability to bind microbial ligands. Ligation of CD180 by monoclonal antibodies leads to B cell activation, upregulation of CD80/CD86, and increase in cell size. CD180 consists of extracellular leucine-rich repeats (LRR) and a short cytoplasmic tail. The extracellular LRR is part of the cell surface receptor complex, RP105/MD-1. Diseases associated with CD180 dysfunction include Kawasaki Disease in children and may play a role in rheumatic disease pathology.
Specifications
| CD180 | |
| Polyclonal | |
| Unconjugated | |
| Cd180 | |
| CD180; CD180 antigen; CD180 molecule; F630107B15; LY64; Ly78; Ly-78; lymphocyte antigen 64; Lymphocyte antigen 78; lymphocyte antigen-64, radioprotective, 105kDa; MGC126233; MGC126234; radioprotective 105 kDa protein; RP105 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4064 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99467 | |
| Cd180 | |
| Recombinant Protein corresonding to Human CD180. Recombinant protein control fragment (Product #RP-102048). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.