Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD180 (RP105) Monoclonal Antibody (MHR73-11), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5012947
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-129-47 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-129-47 Supplier Invitrogen™ Supplier No. 14180982
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The MHR73-11 monoclonal antibody reacts with human CD180 (RP105). This 105 kDa type I transmembrane molecule is a member of the TLR family of proteins characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. RP105 physically associates with another molecule called MD-1 and is expressed on B, monocytes/macrophages, and dendritic cells. Histological studies show that RP105 is expressed mainly on mature B cells in mantle zones, while germinal center cells are either dull or negative. The RP105/MD-1 complex in concert with TLR4 mediates B cell recognition and signaling of LPS. MHR73-11 activates B cells, leading to increases in cell size, expression of the costimulatory molecule CD80, and DNA synthesis. Moreover, ligation of RP105 protects B cells from irradiation- or dexamethasone-induced apoptosis. Thus, RP105 is a signal transduction molecule and plays a role in regulation of B cell growth and death. A significant proportion of circulating B cells in SLE patients is RP105 negative. Loss of RP105 is associated with B cell activation and increased disease activity in SLE patients. Applications Reported: MHR73-11 has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. MHR73-11 has also been reported in in vitro functional studies. (Please use Functional Grade purified MHR73-11, cat. 16-1809, in functional assays.).

CD180 (RP105, Bgp95, LY64) is a type I membrane glycoprotein of Toll-like receptor (TLR) family. CD180 cytoplasmic tail is short and unlike the TLRs, it lacks the TIR domain. CD180 expression is dependent on the coexpression of its helper molecule, MD-1, and mirrors that of TLR4 on antigen-presenting cells. CD180 regulates recognition of LPS and signaling in B cells, via interacting directly with the TLR4 signaling complex, inhibiting its ability to bind microbial ligands. Ligation of CD180 by monoclonal antibodies leads to B cell activation, upregulation of CD80/CD86, and increase in cell size. CD180 consists of extracellular leucine-rich repeats (LRR) and a short cytoplasmic tail. The extracellular LRR is part of the cell surface receptor complex, RP105/MD-1. Diseases associated with CD180 dysfunction include Kawasaki Disease in children and may play a role in rheumatic disease pathology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD180 (RP105)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MHR73-11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd180
Gene Accession No. Q99467
Gene Alias CD180; CD180 antigen; CD180 molecule; F630107B15; LY64; Ly78; Ly-78; lymphocyte antigen 64; Lymphocyte antigen 78; lymphocyte antigen-64, radioprotective, 105kDa; MGC126233; MGC126234; radioprotective 105 kDa protein; RP105
Gene Symbols Cd180
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4064
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.