Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD181 Mouse, anti-Human, R718, Clone: 5A12, BD OptiBuild™

Catalog No. BDB752255 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752255 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752255 Supplier BD Biosciences Supplier No. 752255
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with Interleukin-8 receptor A (IL-8RA), a G protein-linked seven transmembrane domain protein, expressed on monocytes and granulocytes. It is also expressed on a subset of T cells. Both IL-8RA and IL-8RB are high affinity receptors for IL-8. IL-8 is an inflammatory cytokine which induces chemotaxis of neutrophils, basophils and T lymphocytes. IL-8RA and IL-8RB were first clustered as CD128a and CD128b, respectively in the VIIth HLDA workshop, and then were re-clustered as CD181 and CD182, respectively in the VIIIth HLDA workshop.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD181
Applications Flow Cytometry
Classification Monoclonal
Clone 5A12
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.