Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD185 (CXCR5) Monoclonal Antibody (MU5UBEE), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD185 (CXCR5) |
---|---|
Clone | MU5UBEE |
Concentration | 1 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H037T03B08A
![]() |
Invitrogen™
H037T03B08A |
100 Tests |
Each for $430.00
|
|
|||||
H037T02B08A
![]() |
Invitrogen™
H037T02B08A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD185 (CXCR5) Monoclonal antibody specifically detects CD185 (CXCR5) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow CytometrySpecifications
CD185 (CXCR5) | |
1 μg/Test | |
Monoclonal | |
Mouse | |
Human, Monkey, Rhesus Macaque | |
P32302 | |
643, 701792 | |
IgG2b κ | |
Affinity chromatography | |
CXCR5 |
MU5UBEE | |
Flow Cytometry | |
NovaFluor Blue 660-120S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Blr1; Blr-1; Burkitt lymphoma receptor 1; burkitt lymphoma receptor 1 homolog; Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif) receptor 5); Burkitt lymphoma receptor 1, GTP-binding protein; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD185; CD185 antigen; chemochine (C-X-C motif) receptor 5; chemokine (C-X-C motif) receptor 5; CXC; C-X-C chemokine receptor type 5; CXC motif chemokine; C-X-C motif chemokine receptor 5; CXCR5; CXC-R5; CXCR-5; EGK_06966; Fusin; Gpcr6; GPR9; HUMSTR; LESTR; MDR15; MDR-15; Monocyte-derived receptor 15; neurolymphatic receptor; NLR | |
N-terminal extracellular domain. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title