Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Antibody (4G7) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$280.00 - $596.50
Specifications
Antigen | CD19 |
---|---|
Clone | 4G7 |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25266425
![]() |
Novus Biologicals
NBP2526640.025MG |
0.025 mg |
Each for $280.00
|
|
|||||
NBP25266402
![]() |
Novus Biologicals
NBP2526640.2MG |
0.2 mg |
Each for $596.50
|
|
|||||
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, CyTOF-ready.Specifications
CD19 | |
Flow Cytometry 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
CD19 | |
IgG | |
Protein A or G purified | |
Recognises human CD19. |
4G7 | |
Flow Cytometry, Immunofluorescence, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
930 | |
Human CCL (chronic lymphocytic leukemia) cells. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title