Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Antibody (Bu12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | CD19 |
|---|---|
| Clone | Bu12 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:50-1:200, Immunoprecipitation 1:10-1:500 |
| Applications | Immunoprecipitation, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017569
![]() |
Novus Biologicals
NB100643600.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064360
![]() |
Novus Biologicals
NB10064360 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
| CD19 | |
| 1.0 mg/mL | |
| Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| P15391 | |
| 930 | |
| Human EB-4 Burkitt lymphoma cell line. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Bu12 | |
| Flow Cytometry 1:50-1:200, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| CD19 | |
| IgG1 | |
| Protein G purified | |
| NB100-64360 recognizes human CD19, a 95kD cell surface glycoprotein, which is expressed on cells of the B cell lineage and follicular dendritic cells but absent on plasma cells. CD19 is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title